Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8390

Essential Molecular Biology: A Practical Approach, Volume 1, 2nd ed.

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-10:
0-19-963642-7
ISBN-13:
978-0-19-963642-6
UNSPSC Code:
55101509
PubChem Substance ID:

publication info

T.A. Brown, ed., Oxford University Press, 2000, 264 pp., soft cover

manufacturer/tradename

Oxford University Press

General description

There are two books in this series. Volume 1 concentrates on the procedures for DNA and RNA manipulation, purification, electrophoresis and the construction and cloning of recombinant molecules. It also includes a survey of cloning vectors of Escherichia Coli. As with the first edition, the objective is to combine solid practical information with sufficient background material to meet the requirements of any enduser

Table of Contents

Getting Started in Molecular Biology
Microbiological Techniques for Molecular Biology: bacteria and phages
Purification of DNA
Purification of RNA
Nucleic Acid Electrophoresis in Agarose Gels
Recovery of DNA from Electrophoresis Gels
Construction of Recombinant DNA Molecules
Generation and Identification of Recombinant Clones
Survey of Cloning Vectors for Escherichia coli
A1: Important Escherichia coli Strains
A2: Recipes and General Procedures
A3: Restriction Endonucleases

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service