Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8278

Sigma-Aldrich

S1P5 C-Terminal Blocking Peptide

buffered aqueous solution

Synonym(s):

EDG-8, Endothelial Cell Differentiation Gene 8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

form

buffered aqueous solution

Quality Level

mol wt

~2.5 kDa

shipped in

wet ice

storage temp.

−20°C

Physical form

Solution in phosphate buffered saline.

Analysis Note

For use as a control in immunoblotting for specific blocking of Anti-S1P5 (E7528).

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Im et al.
The Journal of biological chemistry, 275(19), 14281-14286 (2000-05-09)
Three G protein-coupled receptors (Edg-1, Edg-3, and Edg-5) for the lysolipid phosphoric acid mediator sphingosine 1-phosphate have been described by molecular cloning. Using a similar sequence that we found in the expressed sequence tag data base, we cloned and characterized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service