Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7905

Sigma-Aldrich

Anti-phospho-Estrogen Receptor-α (pSer167) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-phospho-ER-α (pSer167)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): 3-5 μg/test using extract from MCF-7 breast adenocarcinoma cells (107 cells).
indirect ELISA: 0.1-1.0 μg/mL
western blot: 0.5-2.0 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ESR1(2099)

Immunogen

synthetic phosphopeptide derived from the human region of ER that contains serine 167.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Beena et al.
Veterinary world, 10(8), 964-972 (2017-09-19)
The aim of the present study was to investigate the molecular etiopathology of occurrence of reproductive diseases in female goats. Reproductive diseases in goats account for major economic losses to goat farmers in terms of valuable loss of offspring and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service