Key Documents
E7900
Sigma-Aldrich
Flexible arm electrode holder
Single head
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
manufacturer/tradename
Sigma-Aldrich
General description
Flexible arm to hold and position electrodes in 3-D Space. Base is solid polyethylene and comes with 3 M double-faced mounting tape on the bottom for secure installation. Tygon plastic gently and securely holds probe, ties are also included to secure electrical wires close to the arm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)