Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7772

Sigma-Aldrich

pSE420 Expression vector

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

storage temp.

−20°C

Features and Benefits

4.6-kb supercoiled procaryotic expression vector, offering high-level regulated transcription and enhanced translation of eukaryotic genes in E. coli. The vector contains:
• A strong trc promoter for high-level transcription
• The lacO operator and lacIq repressor for transcriptional regulation
• Translation initiation sequences for optimal expression of mammalian genes in E. coli
• An ampicillin resistance gene for selection
• A colE1 origin for high-copy maintenance in E. coli
• The superlinker (SL2) multiple cloning site.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service