Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7656

Sigma-Aldrich

Endoproteinase Pro-C, recombinant from E. coli

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

powder

specific activity

≥3 units/mg solid

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

One unit corresponds to the amount of enzyme which liberates 1 μmol of p-nitroaninline per minute at pH 7.0 at 25° C, using the substrate Z-Gly-Pro-pNA.

Application

Endoproteinase Pro-C has been used for protein analysis. It acts as a catalyst in the formation of Pro-X bonds in frozen aqueous systems.[1]

Biochem/physiol Actions

Endoproteinase Pro-C catalyzes Pro-Leu bond formation.[1]

Unit Definition

One unit corresponds to the amount of enzyme which liberates 1 μmol of p-nitroaninline per minute at pH 7.0 at 25° C, using the substrate Z-Gly-Pro-pNA.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endoproteinase Pro-C catalyzed Peptide Bond Formation in Frozen Aqueous Systems.
Enz. Microbiol. Technol., 22(7), 617-620 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service