Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7534

Sigma-Aldrich

Anti-Endoglin antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD105, Anti-END, Anti-ENG, Anti-HHT1, Anti-ORW, Anti-ORW1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~95 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysates over-expressing human endoglin

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENG(2022)

General description

Endoglin (CD105) is a membrane glycoprotein that acts as auxillary receptor for TGF-β and is expressed on proliferating vascular endothelial cells. Alternatively spliced isoforms, long endoglin, short endoglin and soluble endoglin are present but the long isoform is predominantly expressed.The major biological functions of endoglin are in angiogenesis and vascular development and homeostasis. It is also vital for wound healing and in development of embryo. Overexpression of endoglin is reported in solid tumors and inflammatory tissues. Expression of endoglin in solid tumors negatively correlates to survival and metastasis and is considered a marker for good prognosis in a variety of solid tumors
Anti-Endoglin specifically recognizes human endoglin (~95 kDa).

Application

Anti-Endoglin antibody may be used for immunoblotting at a working concentration of 1.0-2.0 μg/ml using whole cell lysates of HEK-293T cells over-expressing human endoglin.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolaos A Dallas et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(7), 1931-1937 (2008-04-03)
Endoglin (CD105) is an accessory protein of the transforming growth factor-beta receptor system expressed on vascular endothelial cells. Mutation of the endoglin gene is associated with hereditary hemorrhagic telangiectasias, or Osler-Weber-Rendu syndrome, and has been studied extensively in the context
José M López-Novoa et al.
American journal of physiology. Heart and circulatory physiology, 299(4), H959-H974 (2010-07-27)
Endoglin (CD105) is an integral membrane glycoprotein that serves as a coreceptor for members of the transforming growth factor-β superfamily of proteins. A major role for endoglin in regulating transforming growth factor-β-dependent vascular remodeling and angiogenesis has been postulated based
A Gougos et al.
The Journal of biological chemistry, 265(15), 8361-8364 (1990-05-25)
Endoglin is a major glycoprotein of human vascular endothelium. As observed with monoclonal antibody 44G4, the distribution of endoglin is restricted to endothelial cells in all tissues except bone marrow. cDNA clones were isolated from an endothelial cell lambda gt11

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service