Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7155

Sigma-Aldrich

Endothelin A Receptor preparation human

membrane suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

human

form

membrane suspension

UniProt accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... EDNRA(1909)

Biochem/physiol Actions

Membranes suspended in 50 mM Tris-HCl containing 0.1% Bacitracin, 0.5 mM CaCl2, and 10% sucrose, pH 7.4. Actual concentration and specific binding capacity are provided with each lot.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service