Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7019

Sigma-Aldrich

Exendin-3

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C184H282N50O61S
CAS Number:
Molecular Weight:
4202.57
MDL number:
UNSPSC Code:
51111800

assay

≥97%

Storage temp.

−20°C

Gene Information

human ... GLP1R(2740)

Amino Acid Sequence

His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2

General description

Exendin-3 is a pancreatic secretagogue, which is a member of the glucagon family. It is found in the venom of Heloderma horridurn, which is a Gila monster lizard. It has a molecular mass of 4200Da and is composed of 39 amino acids. 12 amino-acids region in its N-terminal is highly homologous to secretin. Its C-terminal contains an amide group.[1][2]

Application

Exendin-3 has been used to assess its effect in glucagon receptor knockout mice (Gcgr-/-) and wild-type mice, on glucose-stimulated insulin secretion.[3]

Other Notes

Originally isolated from Heloderma horridum (Gila monster lizard) venom.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Eng et al.
The Journal of biological chemistry, 265(33), 20259-20262 (1990-11-25)
An amino-terminal histidyl structure (His1) is characteristic of most peptides in the glucagon superfamily. An assay for His1 peptides performed by amino-terminal amino acid sequencing was used to screen venom from the Gila monster lizard, Heloderma horridum. Two His1 peptides
Actions of Helodermatidae venom peptides and mammalian glucagon-like peptides on gastric chief cells.
Rai, A., et al.
The American Journal of Physiology, 265 (1993)
Bioactive peptides from lizard venoms.
J P Raufman
Regulatory peptides, 61(1), 1-18 (1996-01-16)
Heidi Sørensen et al.
Diabetes, 55(12), 3463-3469 (2006-11-30)
In previous studies, glucagon receptor knockout mice (Gcgr(-/-)) display reduced blood glucose and increased glucose tolerance, with hyperglucagonemia and increased levels of glucagon-like peptide (GLP)-1. However, the role of glucagon receptor signaling for the regulation of islet function and insulin
J Eng et al.
The Journal of biological chemistry, 267(11), 7402-7405 (1992-04-15)
The recent identification in Heloderma horridum venom of exendin-3, a new member of the glucagon superfamily that acts as a pancreatic secretagogue, prompted a search for a similar peptide in Heloderma suspectum venom. An amino acid sequencing assay for peptides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service