Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5657

Sigma-Aldrich

Embedding Molds-Identification Tabs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.47

application(s)

hematology
histology

Quality Level

Storage temp.

room temp

General description

Specimen identification tabs designed for the Peel-A-Way mold system but can be used with any paraffin embedding mold. Each individual tab measures 1 cm (h) × 2 cm (l).

Packaging

Each case contains 8000 tabs.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service