E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate
suitable for electrophoresis, 98.5-101.5%
Synonym(s):
Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2
About This Item
Recommended Products
assay
98.5-101.5%
form
powder
technique(s)
electrophoresis: suitable
impurities
≤0.05% Insoluble matter
mp
248 °C (dec.) (lit.)
solubility
3 M NaOH: 0.16 g/mL, clear, colorless
cation traces
Fe: ≤0.01%
heavy metals (as Pb): ≤50 ppm
storage temp.
room temp
SMILES string
O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
InChI
1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2
Inchi Key
OVBJJZOQPCKUOR-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 3 - STOT RE 2 Inhalation
target_organs
Respiratory Tract
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service