Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5405

Sigma-Aldrich

pTAC-MAT-Tag®-2 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.56

shipped in

dry ice

storage temp.

−20°C

Application

Cytoplasmic expression of C-terminal MAT-Tag® fusion proteins under control of the tac promoter.

Other Notes

Supplied with a pTAC-MAT-2+BAP Control Vector.
Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service