Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5255

Sigma-Aldrich

Estriol 16α-(β-D-glucuronide) sodium salt

≥95% (HPLC)

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol 16-glucuronide, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 16-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C24H31O9Na
CAS Number:
Molecular Weight:
486.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder
assay:
≥95% (HPLC)

biological source

synthetic (organic)

Quality Level

assay

≥95% (HPLC)

form

powder

technique(s)

radioimmunoassay: suitable

solubility

ethanol: water (1:1): 10 mg/mL, clear, colorless

shipped in

ambient

Storage temp.

−20°C

SMILES string

[Na].CC12CCC3C(CCc4cc(O)ccc34)C1CC(OC5OC(C(O)C(O)C5O)C(O)=O)C2O

InChI

1S/C24H32O9.Na.H/c1-24-7-6-13-12-5-3-11(25)8-10(12)2-4-14(13)15(24)9-16(21(24)29)32-23-19(28)17(26)18(27)20(33-23)22(30)31;;/h3,5,8,13-21,23,25-29H,2,4,6-7,9H2,1H3,(H,30,31);;

Inchi Key

BEUFZMWTRVPJQV-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Höller et al.
Acta endocrinologica, 100(1), 57-62 (1982-05-01)
Equimolar concentrations of [4-14C]oestriol and [6,9-3H]oestriol 16 alpha-monoglucuronide were simultaneously perfused through isolated rat livers. Oestriol was hydroxylated to 2-hydroxyoestriol and 6 xi-hydroxyoestriol; 2-hydroxyoestriol was further methylated to 2-methoxyoestriol. Oxidoreduction of oestriol led to the formation of 16 alpha-hydroxyoestrone, 16-0xooestradio-17
K Musch et al.
Zeitschrift fur Geburtshilfe und Perinatologie, 185(5), 284-287 (1981-10-01)
A method based on the inhibition of hemagglutination for the determination of estriol-16-alpha-glucuronide is compared to the method of Kober and Ittrich. A sufficient degree of correlation (p less than 0.001) is found, when the daily estrogen production (Kober-Ittrich) in
W J Brock et al.
The Journal of pharmacology and experimental therapeutics, 229(1), 175-181 (1984-04-01)
The uptake of estradiol-17 beta(beta-D-glucuronide) (E(2)17G), estriol-16 alpha(beta-D-glucuronide (E(3)16G), estradiol-17 beta-3-(beta-D-glucuronide) (E(2)3G) and taurocholate (TC) into hepatocytes isolated from male and female rats was examined and found to be linear for at least 75 sec and to exhibit Michaelis-Menten kinetics.
F Andreolini et al.
Clinical chemistry, 31(1), 124-126 (1985-01-01)
In this relatively simple, rapid assay of estriol-16 alpha-glucuronide in pregnancy urine, the urine sample is diluted 20-fold with phosphate buffer (pH 5.2) containing 360 mL of acetonitrile and 2 g of cetyltrimethylammonium bromide per liter, then directly injected into
T Nambara et al.
Journal of steroid biochemistry, 16(4), 533-538 (1982-04-01)
The preparation and antigenic properties of estriol 16-glucuronide-bovine serum albumin (BSA) and estradiol 17-glucuronide-BSA conjugates in which the hapten molecule is linked to the carrier protein through an (O-carboxymethyl)oxime bridge at the C-6 position on the steroid nucleus, have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service