Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5030

Sigma-Aldrich

pT7-FLAG-MAT-Tag®-2 Expression Vector

Synonym(s):

ddddk tag, dykddddk tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

shipped in

dry ice

storage temp.

−20°C

Application

Cytoplasmic expression of C-terminal FLAG®-MAT-Tag® dual tagged fusion proteins under the T7/lac promoter.

Browse additional application references in our FLAG® Literature portal.

Other Notes

Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG is a trademark of Sigma-Aldrich Co. LLC
pT7-FLAG-MAT is a trademark of Sigma-Aldrich Co. LLC

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bacterial Expression Vectors: T7 Promoter System. T7 Vectors for Highest Expression Levels in Bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service