Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E5029

Sigma-Aldrich

Anti-EphB6 antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

western blot: 1 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ephb6(13848)

General description

The antibody shows less than 1% cross-reactivity with recombinant mouse EphB2, EphB3, and EphB4 in ELISA and immunoblotting assays.

Immunogen

purified recombinant mouse EphB6 extracellular domain.

Biochem/physiol Actions

EPH receptor B6 is a protein-tyrosine kinase family that belongs to the ephrin receptor subfamily. EphB receptors and EphrinB ligands have a regulatory effect on endothelial cell function, angiogenesis, migration, cell growth and survival.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, pH 7.4, with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ombretta Salvucci et al.
Advances in cancer research, 114, 21-57 (2012-05-17)
Eph receptor tyrosine kinases and their Ephrin ligands represent an important signaling system with widespread roles in cell physiology and disease. Receptors and ligands in this family are anchored to the cell surface; thus Eph/Ephrin interactions mainly occur at sites
eph, the largest known family of putative growth factor receptors.
N L Tuzi et al.
British journal of cancer, 69(3), 417-421 (1994-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service