Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E4905

Sigma-Aldrich

pT7-MAT-Tag®-FLAG-2 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

dry ice

storage temp.

−20°C

Application

Cytoplasmic expression of N-terminal MAT-Tag®, C-terminal FLAG® dual tagged fusion proteins under the T7/lac promoter.

Other Notes

Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG-MAT is a trademark of Sigma-Aldrich Co. LLC
pT7-MAT-FLAG is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service