Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E4902

Sigma-Aldrich

Anti-γ-Epithelial Cell Sodium Channel antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-γ-ENaC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 75 kDa

species reactivity

rat

technique(s)

immunoprecipitation (IP): suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCNN1G(6340)
rat ... Scnn1g(24768)

Immunogen

synthetic peptide corresponding to amino acids 630-649 of human γ-ENaC.

Application

Anti-γ-Epithelial Cell Sodium Channel antibody produced in rabbit is suitable for the following applications:
  • Immunoprecipitation
  • Western blotting
  • Immunoblotting

Biochem/physiol Actions

The epithelial sodium channel (ENaC) is expressed in sodium-absorbing epithelia and also found in ocular tissues. It may play an important role in aqueous humor secretion and glaucoma. There are four ENaC subunits (αβγδ) and are expressed in the normal human eye. ENaC activation by proteases may be involved in cystic fibrosis, sodium retention and the pathogenesis of arterial hypertension in the context of renal disease. The γ-subunit of the epithelial sodium channel (SCNN1G) are associated with both mendelian and quantitative effects on blood pressure. It helps in blood pressure determination.

Physical form

Solution in phosphate buffered saline containing 1.0 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noor A S Ismail et al.
European journal of pharmacology, 732, 32-42 (2014-03-25)
Neural precursor cell expressed, developmentally down-regulated protein 4-2 (Nedd4-2) mediates the internalisation / degradation of epithelial Na(+) channel subunits (α-, β- and γ-ENaC). Serum / glucocorticoid inducible kinase 1 (SGK1) and protein kinase A (PKA) both appear to inhibit this
Bettina Krueger et al.
Investigative ophthalmology & visual science, 53(2), 596-604 (2011-12-15)
The epithelial sodium channel (ENaC) is typically expressed in sodium-absorbing epithelia. Several reports suggest that ENaC is also expressed in ocular tissues and may play a role in aqueous humor secretion and glaucoma. However, the precise localization of ENaC in
Cara J Büsst et al.
Hypertension (Dallas, Tex. : 1979), 58(6), 1073-1078 (2011-10-19)
Variants in the gene encoding the γ-subunit of the epithelial sodium channel (SCNN1G) are associated with both Mendelian and quantitative effects on blood pressure. Here, in 4 cohorts of 1611 white European families composed of a total of 8199 individuals
Zhanjun Jia et al.
American journal of physiology. Renal physiology, 299(1), F155-F166 (2010-03-26)
Aldosterone (Aldo) is a major sodium-retaining hormone that reduces renal sodium excretion and also stimulates sodium appetite. In the face of excess Aldo, the sodium-retaining action of this steroid is overridden by an adaptive regulatory mechanism, a phenomenon termed Aldo
Silke Haerteis et al.
Pflugers Archiv : European journal of physiology, 464(4), 353-365 (2012-08-07)
Proteolytic processing of the amiloride-sensitive epithelial sodium channel (ENaC) by serine proteases is known to be important for channel activation. Inappropriate ENaC activation by proteases may contribute to the pathophysiology of cystic fibrosis and could be involved in sodium retention

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service