E4403
Sartorius eLINE® single channel electronic pipettor
volume 5-120 μL, AC/DC input 110 V AC, US 2-pin plug
Synonym(s):
Biohit pipettes, electronic pipettes, pipettor
About This Item
feature
electronic
single-channel
manufacturer/tradename
Sartorius 730041
availability
not available in EU
volume
5-120 μL
General description
The eLINE modes including:
- Forward Pipetting with Optional Cycle
- Reverse Pipetting
- Manual Pipetting
- Post Delivery Mixing
- Multi-Dispensing
- Multi-Aspirating
- Automated Multi-Dispensing
- Sequential-Dispensing
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)