Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E4029

Sigma-Aldrich

Anti-EMMPRIN antibody produced in goat

affinity isolated antibody

Synonym(s):

Anti-CD147, Anti-Extracellular Matrix Metalloproteinase Inducer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BSG(682)

General description

Extracellular matrix metalloproteinase inducer (EMMPRIN) or cluster of differentiation 147 (CD147) is a protein encoded by the BSG gene in humans. It is also known as Basigin (BSG). It is a widely expressed plasma membrane protein that has been implicated in a variety of physiological and pathological activities. Expression of CD147 is significantly higher in ectopic endometrial tissues in patients with endometriosis compared to normal endometrial tissues.

Immunogen

purified recombinant human extracellular matrix metalloproteinase inducer extracellular domain, expressed in NSO cells.

Application

Anti-EMMPRIN antibody produced in goat is suitable for western blotting at a concentration of 0.1-0.2μg/mL.

Biochem/physiol Actions

CD147 functions as an extracellular matrix metalloproteinase inducer, regulates lymphocyte responsiveness, monocarboxylate transporter expression and spermatogenesis. It interacts with cyclophilin family and acts as a signaling receptor to extracellular cyclophilins A and B. EMMPRIN expression in epithelial ovarian cancers (EOC) is higher than in benign ovarian tumors and normal ovaries. It is associated with the pathogenesis of acute coronary syndrome (ACS) and may act as a biomarker for ACS. EMMPRIN plays important roles in tumor progression and sunitinib resistance in renal cell carcinoma (RCC). It could act as a novel target for the treatment of RCC.[1]

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shasha Xu et al.
Clinical and investigative medicine. Medecine clinique et experimentale, 36(6), E282-E289 (2013-12-07)
Leukotriene B4 (LTB4) and extracellular matrix metalloproteinase (EMMPRIN) have been suggested as modulators of atherosclerotic plaque instability. This study sought to evaluate the potential diagnostic implication of LTB4 and EMMPRIN in patients with acute coronary syndrome (ACS). Patients (n=153) who
Vyacheslav Yurchenko et al.
Immunology, 117(3), 301-309 (2006-02-16)
CD147 is a widely expressed plasma membrane protein that has been implicated in a variety of physiological and pathological activities. It is best known for its ability to function as extracellular matrix metalloproteinase inducer (hence the other name for this
Mototaka Sato et al.
PloS one, 8(9), e74313-e74313 (2013-09-28)
Extracellular matrix metalloproteinase inducer (EMMPRIN) has been reported to play crucial roles, including in angiogenesis, in several carcinomas. However, the correlation between EMMPRIN levels and angiogenesis expression profile has not been reported, and the role of EMMPRIN in renal cell
Aihong Jin et al.
Fertility and sterility, 101(6), 1681-1687 (2014-03-26)
To examine the expression of CD147 in 60 human endometriosis lesions and how CD147 regulates migration and apoptosis in human uterine epithelial (HESs) cells. Experimental clinical study and laboratory-based investigation. Hospital and academic research center. Sixty women with chocolate cysts
Sebastian Szubert et al.
Journal of cancer research and clinical oncology, 140(3), 361-369 (2014-01-01)
The primary aim of this paper was to evaluate the expression of extracellular matrix metalloproteinase inducer (EMMPRIN) and its relationship with proangiogenic factors and microvessel density (MVD) in ovarian cancer. The study group included 58 epithelial ovarian cancers (EOCs), 35

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service