Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E3779

Sigma-Aldrich

Anti-Endostatin antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry (frozen sections): 15 μg/mL
indirect ELISA: 0.5-1.0 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Col18a1(12822)

Immunogen

purified recombinant mouse endostatin expressed in Sf21 cells.

Application

Anti-Endostatin antibody produced in goat is suitable for the following applications:
  • Immunohistochemistry (frozen sections) at a concentration of 15μg/mL
  • Indirect ELISA at a concentration of 0.5-1.0μg/mL
  • Western blotting at a concentration of 0.1-0.2μg/mL

Biochem/physiol Actions

Collagen α-1(XVIII) chain is a protein encoded by the COL18A1 gene in humans. It is mapped to chromosome 21q22.3. Endostatin is a 20kDA C-terminal fragment of collagen XVIII that is found in proteoglycan/collagen vessel walls and basement membranes. It helps in inhibiting angiogenesis, tumor growth and also attenuates endothelial cell migration. Endostatin treatment reduces the number of functional blood vessels and the matrix density in the granulation tissue. It is associated with inhibition of chorioallantoic membrane angiogenesis induced by basic fibroblast growth factor (FGF-2) or vascular endothelial growth factor (VEGF).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Dixelius et al.
Cancer research, 62(7), 1944-1947 (2002-04-04)
Endostatin, an endogenous angiogenesis inhibitor, attenuates endothelial cell migration through an unknown mechanism. We show that endostatin induced tyrosine phosphorylation of focal adhesion kinase and paxillin, and promoted formation of focal adhesions and actin stress fibers, similar to fibroblast growth
T Sasaki et al.
Journal of molecular biology, 301(5), 1179-1190 (2000-09-01)
Endostatin is a fragment of the C-terminal domain NC1 of collagen XVIII that inhibits angiogenesis and tumor growth. We report the characterization of a collagen XV endostatin analogue and its parent NC1 domain, obtained by recombinant expression in mammalian cells.
U K Zatterstrom et al.
Cell structure and function, 25(2), 97-101 (2000-07-08)
The angiogenesis inhibitor endostatin is a 20 kDA C-terminal fragment of collagen XVIII, a proteoglycan/collagen found in vessel walls and basement membranes. The endostatin fragment was originally identified in conditioned media from a murine endothelial tumor cell line. Endostatin inhibits
W Bloch et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 14(15), 2373-2376 (2000-10-12)
Endostatin is a cleavage product of collagen XVIII that strongly inhibits tumor angiogenesis. To determine if endostatin affects other angiogenic processes, we generated full-thickness excisional wounds on the back of mice that were systemically treated with recombinant murine endostatin. No
S P Oh et al.
Genomics, 19(3), 494-499 (1994-02-01)
Types XV and XVIII collagen belong to a unique and novel subclass of the collagen superfamily for which we have proposed the name the MULTIPLEXIN family. Members of this class contain polypeptides with multiple triple-helical domains separated and flanked by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service