Recommended Products
conjugate
TRITC conjugate
form
buffered aqueous solution
extent of labeling
1-5 mol fluorochrome per mol protein
technique(s)
immunohistochemistry: 100 μg/mL using human appendix/tonsil sections and Monoclonal Anti Actin, α-Smooth Muscle as primary antibody
indirect immunofluorescence: 1:200
shipped in
dry ice
storage temp.
−20°C
Application
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service