Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E2781

Sigma-Aldrich

Anti-Estrogen-Related Receptor α 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERRα, Anti-ERR1, Anti-NR3B1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~52 kDa (unidentified band is seen at ~37 kDa)

species reactivity

human, rat

technique(s)

western blot: 2 μg/mL using using COS-7 transfected cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ESRRA(2101)

Immunogen

synthetic peptide M(1)SSQVVGIEPL­YIKAE(16) corresponding to amino acid residues 1-16 from human ERR1 protein.

Biochem/physiol Actions

Estrogen-Related Receptors (ERR) are hormone receptors that bind estrogen response elements that in turn bind specific DNA sequences and regulate transcription. Three ERRs are known, alpha, beta and gamma, with highly conserved ligand and DNA binding sequences. ERRα is selectively expressed in kidney, heart, cerebellum, intestine and skeletal muscle. It is an important modulator of energy balance, food intake, neuropeptide levels and response to stress.

Physical form

Provided as ~100 μg of affinity purified antibody (1 mg/mL) in phosphate buffered saline containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angie L Bookout et al.
Cell, 126(4), 789-799 (2006-08-23)
In multicellular organisms, the ability to regulate reproduction, development, and nutrient utilization coincided with the evolution of nuclear receptors (NRs), transcription factors that utilize lipophilic ligands to mediate their function. Studying the expression profile of NRs offers a simple, powerful
Mardi S Byerly et al.
BMC physiology, 13, 10-10 (2013-09-24)
Estrogen-related receptors (ERRs) are orphan nuclear hormone receptors expressed in metabolically active tissues and modulate numerous homeostatic processes. ERRs do not bind the ligand estrogen, but they are able to bind the estrogen response element (ERE) embedded within the ERR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service