Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E2654

Sigma-Aldrich

elF-4E human

Synonym(s):

Eukaryotic initiation factor 4E

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

packaging

vial of 1 μg

Quality Level

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

eIF-4E or eukaryotic Initiation Factor 4E, plays a critical role in the initiation of the protein synthesis process. The structure recognizes and binds to the 7-methylguanosine-containing mRNA ′cap′.

Physical form

Supplied as 1 μg of protein per vial in 50 μl of 50 mM Tris-HCl, pH 7.0, containing 14 mM 2-Mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF, 1 mM EDTA, and 10% glycerol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service