E2033
14-3-3 Epsilon histidine-tagged human
recombinant, expressed in E. coli, C-terminal, ≥90% (SDS-PAGE), buffered aqueous solution
Synonym(s):
14-3-3ε, FLJ45465, KCIP-1, MDCR, MDS, Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide, YWHAE
About This Item
recombinant
expressed in E. coli
assay
≥90% (SDS-PAGE)
form
buffered aqueous solution
UniProt accession no.
shipped in
dry ice
Storage temp.
−20°C
Gene Information
human ... YWHAE(7531)
General description
Biochem/physiol Actions
Physical form
related product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service