Key Documents
E1888
UV blocking eye glasses
with side shields
About This Item
UNSPSC Code:
46181802
Product E1888 is not currently sold in your country. Contact Technical Service
availability
not available in EU
General description
Clear plastic glasses with side shields block UV radiation below 400 nm. For full-face protection against "sunburn," we recommend the UV-blocking face shield.
Choose from one of the most recent versions:
Certificates of Analysis (COA)