E1783
Monoclonal Anti-eIF5A antibody produced in mouse
~1.0 mg/mL, clone IF5-88, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-Eukaryotic translation initiation factor 5A, Anti-eIF-4D, Anti-eIF-5A-1, Anti-eIF-5A1
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
IF5-88, monoclonal
form
buffered aqueous solution
mol wt
antigen ~17 kDa
species reactivity
canine, monkey, mouse, rat, human
concentration
~1.0 mg/mL
technique(s)
immunocytochemistry: suitable
western blot: 1-2 μg/mL using 3T3 total cell extract
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... EIF5A(1984)
mouse ... Eif5a(276770)
rat ... Eif5a(287444)
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
related product
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service