Key Documents
E0389
Sigma-Aldrich
Electrophoresis film for polyacrylamide gels
7 mil (0.2 mm) thickness
About This Item
UNSPSC Code:
41100000
NACRES:
NB.31
material
polyester
Application
Electrophoresis film is a flexible, transparent polyester film specifically designed to support polyacrylamide gels. Ideal for casting horizontal IEF gels. One side has been treated with a hydrophilic coating which enhances adhesion with aqueous polyacrylamide gel media. The acrylamide monomers covalently attach to the coating during polymerization. Gels remain attached during electrophoresis, staining and drying.
Other Notes
124 mm × 258 mm sheets
Choose from one of the most recent versions:
Certificates of Analysis (COA)