Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E0160

Sigma-Aldrich

Estrogen Receptor human

membrane suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

(breast carcinoma cell membranes)

form

membrane suspension

concentration

19.8 mg/mL protein

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

Membranes are suspended in 25 mM TRIS-HCl (pH 7.4 at 25°C) containing 1.5 mM EDTA, 1 mM DTT and 25 mM Na molybdate.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service