Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

E005134

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt dihydrate

suitable for electrophoresis, 99.0-101.0% (titration)

Synonym(s):

Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein/REAXYS Number:
3900609
EC Number:
UNSPSC Code:
12352200

grade

Molecular Biology

assay

99.0-101.0% (titration)

form

powder

availability

available only in China

technique(s)

electrophoresis: suitable

impurities

≤0.005% Insolubles
≤0.1% Nitrilotriacetic acid (NTA)

mp

248 °C (dec.) (lit.)

cation traces

Fe: ≤0.005%
Pb: ≤0.002%

foreign activity

DNase, RNase, NICKase, and protease, none detected

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

Inchi Key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Pictograms

Exclamation markHealth hazard

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - STOT RE 2 Inhalation

target_organs

Respiratory Tract

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service