DUO80102
Duolink® In Situ Brightfield Mounting Medium
About This Item
Recommended Products
product line
Duolink®
technique(s)
proximity ligation assay: suitable
storage temp.
20-25°C
Application
To perform a complete Duolink in situ experiment you will need two primary antibodies (IHC or IF validated) that recognize two target epitopes. Additional reagents required include a pair of PLA probes, one PLUS and one MINUS , your choice of Detection Reagents. Optional reagents include Wash Buffers and Mounting Medium.
Analysis is carried out using standard immunofluorescence assay equipment. HRP/Novared is also available for bright field detection. Quick and reliable quantification can be performed using the Duolink Image Tool.
Application Note
Two primary antibodies raised in different species are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF) or immunohistochemistry (IHC) assay to determine the optimal fixation, blocking, and titer conditions.
Duolink in situ reagents are suitable for use on fixed cells, cyt-spin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.
Find answers to commonly asked question on our Duolink FAQ page
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
target_organs
hearing organs, Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
73.4 °F - closed cup
flash_point_c
23 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Applications to detect, quantify and visualize protein-protein interactions, post-translational modifications and low expression protein detection using proximity ligation assay
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service