Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DOPA1003

Sigma-Aldrich

Dopaminergic neurons, PARK7 (-/-) KO Line

IPSC derived

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

growth mode

Adherent

karyotype

2n=46

morphology

elongated/polarized, neuronal

relevant disease(s)

Parkinson′s disease

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

biallelic knock-out of PARK7 gene

Application

These products provide unique and versatile tools for the study of central nervous system (CNS) disorders, such as Alzheimer′s disease (AD), Parkinson′s disease (PD), autism, schizophrenia, and amyotrophic lateral sclerosis (ALS), as well as for drug screening and toxicology applications in the neural space.

Features and Benefits

This product contains cryo-preserved, pre-differentiated dopaminergic neurons derived from a footprint-free, karyotype normal human iPSC line with a homozygous knockout in PARK7 made using zinc fingers. It is designed for customers to generate mature dopaminergic neurons using the optimized maturation medium and supplements (DOPA1050, DOPA1051 sold separately). Mature and functional dopaminergic neurons can be obtained within 12-14 days. The dopaminergic precursors can be seeded on various culture vessel formats including 96-well plates on either glass or plastic surfaces and cultured as adherent cells. Shortly after seeding, the cells proliferate slightly for up to 4 days and show extensive neurite outgrowth and proper neuronal morphology. In general, at 12-14 days post-seeding, the cell population will contain >80% neurons, >30% TH+ dopaminergic neurons and < 15% GFAP+ astrocytes.

Culture Medium

User must also purchase Dopa Maturation Medium, DOPA1050 (50mL) or DOPA1051 (250mL). Sold separately.

Subculture Routine

No subculture routine is necessary. See technical bulletin for additional details.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej Swistowski et al.
Stem cells (Dayton, Ohio), 28(10), 1893-1904 (2010-08-18)
Human induced pluripotent stem cells (iPSCs) reprogrammed from somatic cells represent a promising unlimited cell source for generating patient-specific cells for biomedical research and personalized medicine. As a first step, critical to clinical applications, we attempted to develop defined culture
Yiping Yan et al.
Stem cells translational medicine, 2(11), 862-870 (2013-10-12)
Human pluripotent stem cells (hPSCs), including human embryonic stem cells and human induced pluripotent stem cells, are unique cell sources for disease modeling, drug discovery screens, and cell therapy applications. The first step in producing neural lineages from hPSCs is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service