Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

DNB400

Sigma-Aldrich

GenElute FFPE DNA Purification Kit

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600
NACRES:
NA.52

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

Sigma′s GenElute FFPE DNA Purification Kit provides a rapid method for the isolation and purification of genomic DNA from formalin-fixed paraffin-embedded (FFPE) tissue samples. Using formalin to fix tissues leads to crosslinking of the nucleic acids and proteins, and the process of embedding the tissue samples can also lead to fragmentation of the nucleic acids over time. Sigma′s GenElute FFPE DNA Purification Kit provides conditions that allow for the partial reversing of the formalin modifications, resulting in a high quality and yield of nucleic acids. The DNA is purified from other cellular components without the use of phenol or chloroform. The purified genomic DNA is of the highest integrity, and can be used in a number of downstream applications including qPCR, mutation screening, microarray analyses, sequencing, Southern blotting and SNP analysis.

  • Fast and easy processing using rapid spin-column format
  • High yields of gDNA
  • No phenol or chloroform extractions

Application

  • PCR and Quantitative PCR
  • Mutation screening
  • Sequuencing
  • Southern blotting
  • Microarrays
  • SNP analysis

Features and Benefits

  • High quality and integrity of the isolated DNA - The purified DNA is of the highest quality and integrity, and can be used in any sensitive downstream applications.
  • High yields - Norgen′s FFPE DNA Purification Kit allows for the purification of high yields of DNA.
  • No phenol:chloroform extractions - DNA is isolated from FFPE tissue samples without the use of harmful chemicals such as phenol or chloroform.
  • Rapid procedure - Isolate DNA from FFPE tissue sections using a rapid spin column format in as little as 1 hour

Unit Definition

Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service