Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354852

Sigma-Aldrich

Corning® TransportoCells Organic Cation Transporter 1 (OCT1 / SLC22A1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

shelf life

3 yr (In liquid nitrogen vapor.)

shipped in

dry ice

storage temp.

−196°C

General description

Corning TransportoCells products are high performance mammalian cells in a convenient, cryopreserved format that transiently overexpress a single human SLC transporter protein. The frozen cells deliver robust data, while eliminating the time required to culture and maintain stable cell lines.

Culturing and maintaining stable transporter cell lines can be expensive and time-consuming. In addition, it can take a week or more to prepare cells for assaying. Corning TransportoCells products can be thawed, plated and assayed in just two days with high uptake ratios.

Application

Convenient and Cost-efficient
Corning TransportoCells products are a convenient, cost-efficient alternative to maintaining stable cell lines. They provide the utmost flexibility for experimental planning. The cells can be removed from storage one day and assayed the following day.

Robust and Validated
The model has been fully validated for substrate specificity, transporter kinetics, and inhibition profiles to ensure data are consistent with existing transporter cell models.

Supports Regulatory Recommendations
Corning TransportoCells products are a convenient, cost-efficient alternative to maintaining stable cell lines. They provide the utmost flexibility for experimental planning. The cells can be removed from storage one day and assayed the following day.

Features and Benefits

  • Cryopreserved format provides flexibility for experimental planning.
  • Cells are readily available, can be stored onsite and shipped globally.
  • Cells can be thawed, plated and assayed in just two days.
  • Robust results with uptake ratios ≥10 fold.
  • Consistent with other mammalian cell-based models.
  • One vial contains 10 million cells and supports one 24-well or one 96-well plate.
  • Includes all current USFDA and European Medicines Agency (EMA)-recommended SLC drug transporters.

Legal Information

Corning is a registered trademark of Corning, Inc.
TransportoCells is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service