Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9894

SAFC

Diethyl chlorophosphite

Synonym(s):

Ethyl phosphorochloridite

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

storage temp.

2-8°C

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

33.8 °F - closed cup

flash_point_c

1 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service