Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9191

Sigma-Aldrich

JumpStart Taq ReadyMix with dUTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
Product D9191 is not currently sold in your country. Contact Technical Service

concentration

2.5 units/reaction (50 μL reaction volume)

shipped in

wet ice

storage temp.

−20°C

Features and Benefits

  • The ideal ReadyMix for high throughput, quantitative PCR applications where carryover contamination is a concern
  • The hot start mechanism using JumpStart Taq antibody prevents non-specific product formation; assembled PCR reactions can remain at room temperature for up to 2 hours without compromising performance
  • The incorporation of dUTP provides the ability to eliminate contaminating PCR products from earlier reactions with uracil-DNA glycosylase; prevents carryover contamination

Other Notes

JumpStart Taq ReadyMix with dUTP for Quantitative PCR Kit provides the performance enhancements of our JumpStart Taq Antibody for hot start in a convenient, easy-to-use reaction mixture. The mix has no added dyes, making it ideal for performing high throughput quantitative PCR (qPCR) methods that rely on a fluorescent probe.

Principle

The ReadyMix has incorporated dUTP in place of TTP to facilitate carryover prevention. This nucleotide mixture provides the ability to eliminate contaminating PCR products by use of a Uracil-DNA glycosylase (UNG). The ready-to-use mixture of JumpStart Taq DNA polymerase, 99% pure deoxynucleotides and reaction buffer is provided in a 2× concentrate. Simply add 25 μL of the 2× mix to the DNA template, primers, fluorescent probe, UNG, and water. At room temperature, the JumpStart Taq antibody inactivates the DNA polymerase. However, during the first denaturation step of the cycling process, the complex dissociates and the polymerase becomes fully active.

Legal Information

JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • R4526Reference Dye for Quantitative PCR, 100 ×, solutionSDS

  • M8787Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCRSDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service