Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9019

Sigma-Aldrich

Distearoylglycerol mixed isomers

≥99%

Synonym(s):

DG(18:0/18:0/0:0)[rac]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥99%

form

powder

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3

InChI key

UHUSDOQQWJGJQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Approx. 50% 1,3- and 50% 1,2-isomer.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service