Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D7939

Sigma-Aldrich

Cyclic 3-deaza-adenosine diphosphate-ribose

~95% (HPLC), oil

Synonym(s):

3-Deaza-cADP-ribose, 3-Deaza-cyclic adenosine 5′-diphosphate ribose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N4O13P2
Molecular Weight:
540.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~95% (HPLC)

form

oil

color

light yellow

solubility

H2O: freely soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

Nc1nccc2n(cnc12)[C@@H]3O[C@@H]4COP(=O)(O[C@H]4[C@H]3O)OP(O)(=O)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H]5O

InChI

1S/C16H22N4O13P2/c17-14-9-6(1-2-18-14)20(5-19-9)15-12(24)13-8(29-15)4-28-35(27,31-13)33-34(25,26)32-16-11(23)10(22)7(3-21)30-16/h1-2,5,7-8,10-13,15-16,21-24H,3-4H2,(H2,17,18)(H,25,26)/t7-,8-,10-,11-,12-,13-,15-,16+,35?/m1/s1

InChI key

OQRDOVWETUSZCW-NDRVCTFCSA-N

Biochem/physiol Actions

Heat and hydrolysis resistant cyclic ADP ribose agonist.

Legal Information

US Patent No. 6,593,307

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service