DPO-1 is an inhibitor of human Kv1.5 potassium channel; representative blocker of a novel pharmacophore.
DPO-1 is an inhibitor of human Kv1.5 potassium channel; representative blocker of a novel pharmacophore. The Kv1.5 potassium channel, which underlies the ultrarapid delayed rectifier current, IKur, is reported to be enriched in human atrium versus ventricle, and has been proposed as a target for novel atrial antiarrhythmic therapy. The administration of the IKur blocker (2-isopropyl-5-methyl-cyclohexyl) diphenylphosphine oxide (DPO-1) increases myocardial refractoriness in both atrium and ventricle of rat, but produces an atrial-selective increase in refractoriness in primates; appears to be 15-fold more selective for Kv1.5 vs Kv3.1 channels expressed in Xenopus oocytes. IC50 = 0.16-0.76 μM at 0.1 Hz pulsing frequency; Kd = 0.6 μM.
Features and Benefits
This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
European journal of pharmacology, 849, 59-66 (2019-02-05)
In the present study, we investigated the inhibitory effect of tacrolimus, a macrolide immunosuppressive drug that acts through calcineurin inhibition, on voltage-gated K+ (Kv) channels expressed in native smooth muscle cells isolated from the coronary arteries of rabbits. Tacrolimus reduced
Diphenyl phosphine oxide-1 (DPO-1) is a potent Kv1.5 channel inhibitor that has therapeutic potential for the treatment of atrial fibrillation. Many other Kv1.5 channel blockers also potently inhibit the Kv1.3 channel, but whether DPO-1 blocks Kv1.3 channels has not been
British journal of pharmacology, 177(5), 1164-1186 (2019-10-29)
BK channels play important roles in various physiological and pathophysiological processes and thus have been the target of several drug development programmes focused on creating new efficacious BK channel openers, such as the GoSlo-SR compounds. However, the effect of GoSlo-SR
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 21(4), 415-421 (2017-07-15)
We investigated the inhibitory effect of escitalopram, a selective serotonin reuptake inhibitor (SSRI), on voltage-dependent K
Voltage-gated ion channels (VGCs) are prime targets for the pharmaceutical industry, but drug profiling on VGCs is challenging, since drug interactions are confined to specific conformational channel states mediated by changes in transmembrane potential. Here we combined various optogenetic tools
We offer many products related to potassium channels for your research needs.
Questions
Reviews
★★★★★ No rating value