Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D7319

Sigma-Aldrich

DDR2 (467-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

NTRK R3, TKT, TYRO10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

41-55 nmol/min·mg

mol wt

~70 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DDR2(4921)

Biochem/physiol Actions

DDR2 is a member of a novel subclass of RTKs containing a distinct extracellular region encompassing a factor VIII-like domain and is thought to be involved in the regulation of cell growth, differentiation, and metabolism. DDR2 plays a role in the regulation of collagen turnover mediated by smooth muscle cells in obstructive diseases of blood vessels and the lung.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Ferri et al.
The American journal of pathology, 164(5), 1575-1585 (2004-04-28)
Obstructive diseases of blood vessels and the lung are characterized by degradation and synthesis of new extracellular matrix (ECM) components. Regulated remodeling of the ECM in diseases such as atherosclerosis and lymphangioleiomyomatosis (LAM), both characterized by excessive accumulation of smooth
T Karn et al.
Oncogene, 8(12), 3433-3440 (1993-12-01)
Using a polymerase chain reaction-mediated approach we have characterized cDNAs from human and mouse origin representing a novel type of receptor protein tyrosine kinase (RTK). The deduced amino acid sequence (855 amino acids) of the longest open reading frame has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service