Recommended Products
form
liquid
shipped in
wet ice
Storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
For routine PCR amplifications
Components
Each set includes:
- Taq DNA polymerase supplied at 5 units/μl
- Optimized 10× Reaction Buffer available with or without MgCl2. A separate tube of 25 mM MgCl2 is included with the 10× buffer not containing MgCl2.
- Ultrapure Deoxynucleotide mix containing 10 mM each of ultrapure dATP, dCTP, dGTP and dTTP in molecular biology grade water. It is suitable for use in routine and specialized (long and accurate) PCR, manual and automated sequencing, RT-PCR and cDNA synthesis reactions, DNA labeling and mutagenesis reactions.
Other Notes
Taq SuperPaks are conveniently packaged to include all reagents needed for PCR except for specific primers, template and water. Also available with REDTaq® DNA Polymerase.
Unit Definition
One unit will incorporate 10 nmol of total dNTPs into acid-precipitable DNA in 30 min at 74 °C.
Legal Information
No license is conveyed with the purchase of this product under any of US Patents Nos. 5,804,375, 5,994,056, 6,171,785, 6,214,979, 5,538,848, 5,723,591, 5,876,930, 6,030,787, and 6,258,569, and corresponding patents outside the United States, or any other patents or patent applications, relating to the 5′ Nuclease and dsDNA-Binding Dye Processes. For further information contact the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
SuperPak is a trademark of Sigma-Aldrich Co. LLC
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service