Recommended Products
form
liquid
concentration
5 units/μL
shipped in
wet ice
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Features and Benefits
- KlenTaq LA DNA Polymerase has increased thermostability and processivity, resulting in increased yields
- Amplify difficult structure or GC-rich templates. The increased thermostability allows higher temperature conditions to disrupt difficult secondary structures
- Increased fidelity at up to 4× higher than that of Taq DNA polymerase
- Tolerance to a broad range of magnesium concentrations eliminates the need to optimize MgCl2
- Amplify up to 5 kb genomic targets and up to 20 kb on less complex targets, such as lambda DNA
Packaging
Other Notes
Unit Definition
Legal Information
comparable product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service