Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D4660

Sigma-Aldrich

1-Dehydrotestosterone undecylenate

Synonym(s):

1,4-Androstadien-17β-ol-3-one 17-(10-undecenoate), 17β-Hydroxy-1,4-androstadien-3-one 17-(10-undecenoate)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41100000
NACRES:
NA.24
Pricing and availability is not currently available.

form

solid

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service