Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

D4197

Sigma-Aldrich

SigMatrix Serum Diluent

For in-vitro Diagnostic Use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

human

recombinant

expressed in rice

form

solution

concentration

18-22 mg/mL Biuret

pH

7.2-7.6

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

General description

For more information on other Sigmatrix products visit the Sigmatrix Selection Guide.
SigMatrix is a serum diluent that utilizes recombinant human albumin as the source of the albumin blocking reagent. The use of human albumin expressed in rice can eliminate the source of interfering analytes typically found in human and bovine-derived diluents. Many commercially-available diluents rely on charcoal-stripping which often only results in limited reduction of interfering analytes.

Suitability

Recommended for use in ELISA and LC-MS-based diagnostic applications.

Physical form

SigMatrix Serum Diluent is an aqueous solution containing 2% recombinant human albumin (expressed in rice) in phosphate buffered saline (pH 7.4) with 0.1% ProClin 950.

Legal Information

ProClin is a trademark of LANXESS Corp.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia S Capano et al.
Cell stem cell, 29(6), 918-932 (2022-06-07)
Tau is a microtubule-binding protein expressed in neurons, and the equal ratios between 4-repeat (4R) and 3-repeat (3R) isoforms are maintained in normal adult brain function. Dysregulation of 3R:4R ratio causes tauopathy, and human neurons that recapitulate tau isoforms in

Questions

  1. Is there a replacement available for the discontinued product D4197 in a 100ml size?

    1 answer
    1. There are two other SigMatrix Serum Diluent products available, each with a different regulatory status. D4197 has a regulatory status of 600, D5322 has a regulatory status of 200, and D5197 has a regulatory status of 500. A table on the M Clarity page describes the differences between these regulatory quality levels. All three part numbers appear to be recombinant human albumin. For more information, you can visit the M Clarity page here. Additionally, there are separate links for products D5322 and D5197:

      D5322
      D5197

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service