Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D3688

Sigma-Aldrich

Anti-DR3, Extracellular Domain antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Apo-3, Anti-Death Receptor 3, Anti-LARD, Anti-TRAMP, Anti-Wsl-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human

technique(s)

western blot: 0.5 μg/mL using human Jurkat cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TNFRSF25(8718)

General description

DR3 is a member of the TNF superfamily. It is expressed in lymphocyte rich tissues such as blood, thymus and spleen.

Immunogen

synthetic peptide corresponding to amino acids 59-77 of the extracellular domain of human DR3 precursor.

Application

Anti-DR3 antibody produced in rabbit is suitable for immunoblotting at a working concentration of 0.5μg/mL using total cell lysates from Jurkat cells. It was used as a primary antibody in immunoblotting analysis of adenocarcinoma cells to examine 14-3-3 isoforms expressed in normal and cancerous lung tissues.

Biochem/physiol Actions

DR3 is a death domain-containing receptor that is involved in inducing apoptosis or programmed cell death in cells. Binding of ligand to these receptors sends signals that activate members of the caspase family of proteases, which cause the degradation of chromosomal DNA by activating DNase. It also activates NF-κB, a protein complex that controls transcription of DNA.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqing Qi et al.
International journal of cancer, 113(3), 359-363 (2004-09-30)
14-3-3 Proteins play important roles in a wide range of vital regulatory processes, including signal transduction, apoptosis, cell cycle progression and DNA replication. In mammalian cells, 7 14-3-3 isoforms (beta, gamma, epsilon, eta, sigma, theta and zeta) have been identified
A Singh et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 18(7), 439-450 (1998-08-26)
Apoptotic signals are transduced by five death domain-containing receptors--TNFR1, Fas, DR3, DR4, and DR5--by binding to their ligands. The intracellular portion of all these receptors contains a region, approximately 80 amino acids long, referred to as the "death domain" (DD).
A M Chinnaiyan et al.
Science (New York, N.Y.), 274(5289), 990-992 (1996-11-08)
Tumor necrosis factor receptor-1 (TNFR-1) and CD95 (also called Fas or APO-1) are cytokine receptors that engage the apoptosis pathway through a region of intracellular homology, designated the "death domain." Another death domain-containing member of the TNFR family, death receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service