Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2889

Sigma-Aldrich

1,3-Dimethyluric acid

Synonym(s):

1,3-Dimethyl-2,6,8-trihydroxypurine, 1,3-Dimethyl-7,9-dihydro-1H-purine-2,6,8(3H)-trione, Oxytheophylline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O3
CAS Number:
Molecular Weight:
196.16
Beilstein/REAXYS Number:
193698
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

assay

≥98% (TLC)

form

powder

mp

≥300 °C (lit.)

solubility

1 M NH4OH: soluble 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CN1C(=O)N(C)C2=C(NC(=O)N2)C1=O

InChI

1S/C7H8N4O3/c1-10-4-3(8-6(13)9-4)5(12)11(2)7(10)14/h1-2H3,(H2,8,9,13)

InChI key

OTSBKHHWSQYEHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Mizuki et al.
Antimicrobial agents and chemotherapy, 40(8), 1875-1880 (1996-08-01)
Enoxacin, an antimicrobial fluoroquinolone with a 7-piperazinyl-1, 8-naphthyridine skeleton, is a potent inhibitor of cytochrome P-450-mediated theophylline metabolism. The present study was designed to clarify, using seven enoxacin derivatives, the molecular characteristics of the fluoroquinolone responsible for the inhibition. Three
Andreas Daiber et al.
International journal of molecular sciences, 14(4), 7542-7570 (2013-04-10)
The reaction product of nitric oxide and superoxide, peroxynitrite, is a potent biological oxidant. The most important oxidative protein modifications described for peroxynitrite are cysteine-thiol oxidation and tyrosine nitration. We have previously demonstrated that intrinsic heme-thiolate (P450)-dependent enzymatic catalysis increases
Yu Chul Kim et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 26(1), 114-123 (2005-06-30)
Pharmacokinetic parameters of theophylline and one of its metabolites, 1,3-dimethyluric acid (1,3-DMU), were compared after intravenous and oral administration of aminophylline, 5mg/kg as theophylline, to diabetes mellitus rats induced by alloxan (DMIA) or streptozotocin (DMIS), and their respective control rats.
A O El-Kadi et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(9), 1112-1120 (2000-08-19)
Serum of rabbits with a turpentine-induced acute inflammatory reaction (RS(INFLA)) and serum of humans with a viral infection (HS(INF)) were previously shown to diminish hepatic cytochrome P450 (P450) content and activity. To document the role of reactive oxygen intermediates in
Su Yeon Yu et al.
The Journal of pharmacy and pharmacology, 54(12), 1687-1692 (2003-01-25)
In rats with acute renal failure induced by uranyl nitrate, the hepatic microsomal cytochrome P450 (CYP) 2E1 and CYP3A23 increased 2-4- and 4-times, respectively, CYP2C11 decreased to 80% of control, but the levels of CYP1A2 and CYP2B1/2 were not changed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service