Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2566

Sigma-Aldrich

Monoclonal Anti-DC-SIGN2 antibody produced in mouse

clone 120604, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CD209L, Anti-DC-SIGNR, Anti-Dendritic Cell-Specific ICAM-3 Grabbing Non-Integrin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

120604, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC4M(10332)

Specificity

Recognizes human DC-SIGN2 on transfected cells. The antibody does not react with parental mouse cells and irrevelent transfectants, such as human DC-SIGN1 or dendritic cells.

Immunogen

purified from NIH/3T3 cells transfected with recombinant human DC-SIGN2.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service