D2441
DcR3/Fc Chimera human
>90% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder
Synonym(s):
Decoy Receptor 3, M68, TR6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
recombinant
expressed in baculovirus infected Sf21 cells
assay
>90% (SDS-PAGE)
form
lyophilized powder
mol wt
glycosylated form 60-65 kDa by SDS-PAGE (reducing)
calculated mol wt 57 kDa
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... TNFRSF6B(8771)
Biochem/physiol Actions
DcR3 is a disulfide-linked homodimeric protein that is a member of the TNF receptor family. DcR3 modulates ligands that induce apoptosis.
Disulfide-linked homodimeric protein that is a member of the TNF receptor family; modulates ligands that induce apoptosis.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.
Analysis Note
Biological activity is measured by its ability to inhibit FAS-ligand induced apoptosis of Jurkat cells.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service