Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0286

Sigma-Aldrich

13,14-Dihydro-15-ketoprostaglandin E2 solution

~90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

assay

~90%

form

solution

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCC(=O)CC[C@H]1[C@H](O)CC(=O)[C@@H]1C\C=C/CCCC(O)=O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,16-17,19,23H,2-3,5-6,8-14H2,1H3,(H,24,25)/b7-4-/t16-,17-,19-/m1/s1

InChI key

CUJMXIQZWPZMNQ-XYYGWQPLSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Brennand et al.
Prostaglandins, leukotrienes, and essential fatty acids, 58(5), 369-375 (1998-08-05)
This study aims to investigate potential mechanisms involved in the stimulatory effect of amniotic fluid on prostaglandin production by fetal membranes. A cell culture study of amnion and chorion was obtained following elective caesarean section, incubated with amniotic fluid collected
N Nishigaki et al.
Molecular pharmacology, 50(4), 1031-1037 (1996-10-01)
There are at least four subtypes of prostaglandin E (PGE) receptors. The EP1 and EP3 receptors are coupled to Ca2+ mobilization and the inhibition of adenylate cyclase, respectively, and the EP2 and EP4 receptors are coupled to the same signal
S Sakuma et al.
Prostaglandins, leukotrienes, and essential fatty acids, 51(6), 425-430 (1994-12-01)
The effect of 13-hydroperoxyoctadecadienoic acid (13-HPODE), a hydroperoxy adduct of linoleic acid (LA), on the activities of prostaglandin (PG) synthesizing and catabolizing enzymes in rabbit gastric antral mucosa was examined. 13-HPODE had no effect on the synthesis of PGE2, PGF2
Patricia A Thompson et al.
Cancer prevention research (Philadelphia, Pa.), 3(1), 101-107 (2010-01-07)
Regular use of nonsteroidal anti-inflammatory drugs (NSAID) has been associated with reduced risk of breast cancer. Sulindac, a nonselective NSAID with both cyclooxygenase-2-dependent and -independent activities, is a candidate for breast chemoprevention. We conducted a phase Ib trial in 30
B A Peskar et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 42(3), 327-331 (1991-09-01)
We have demonstrated recently the formation of a biologically active metabolite of prostaglandin (PG) E1, 13,14-dihydro-PGE1, during intravenous infusions of PGE1 in patients with peripheral arterial occlusive disease. We have now investigated the levels of the immediate precursor of 13,14-dihydro-PGE1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service