Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0262

Sigma-Aldrich

1,3-Dimethyluracil

Synonym(s):

1,3-Dimethyluracil, 2,4-Dihydroxy-1,3-dimethylpyrimidine, 1,3-Dimethyl-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
Beilstein/REAXYS Number:
124074
EC Number:
MDL number:
UNSPSC Code:
12352204

mp

119-122 °C (lit.)

SMILES string

CN1C=CC(=O)N(C)C1=O

InChI

1S/C6H8N2O2/c1-7-4-3-5(9)8(2)6(7)10/h3-4H,1-2H3

InChI key

JSDBKAHWADVXFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service