Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D017

Sigma-Aldrich

m-Tyramine hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

3-Hydroxyphenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO · HCl
CAS Number:
Molecular Weight:
173.64
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

off-white to tan

solubility

H2O: ≥10 mg/mL

SMILES string

Cl[H].NCCc1cccc(O)c1

InChI

1S/C8H11NO.ClH/c9-5-4-7-2-1-3-8(10)6-7;/h1-3,6,10H,4-5,9H2;1H

InChI key

GTIWCKXKQGMMQZ-UHFFFAOYSA-N

Biochem/physiol Actions

Dopamine receptor agonist; biological precursor to dopamine.

Caution

Photosensitive

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P L Walker et al.
Annals of clinical biochemistry, 25 ( Pt 3), 304-309 (1988-05-01)
A simple, reliable method for the analysis of urinary meta- and para-tyramine has been developed. Sample purification was achieved by a weak anion-exchange column, followed by extraction into ethyl acetate at pH 10.2. The heptafluorobutyryl derivative was measured by packed
S C Yeung et al.
General pharmacology, 16(5), 517-519 (1985-01-01)
The concentration of homovanillic acid and p-tyramine in the female corpus striatum were increased after ovariectomy. The increases were reversed by chronic administration of estradiol benzoate and potentiated by progesterone. It is proposed that these changes are the consequence of
D A Durden et al.
Neurochemical research, 18(9), 995-1002 (1993-09-01)
Using a new ultrasensitive method the trace biogenic amines, phenylethylamine, meta-tyramine and para-tyramine have been quantitated in brain regions obtained from a single rat. Phenylethylamine concentrations in ng/g wet tissue (mean +/- std. error) were as follows: caudate 2.71 +/-
P S McQuade et al.
European journal of pharmacology, 83(3-4), 277-282 (1982-09-24)
The concentrations of p-tyramine (p-TA), m-tyramine (m-TA), dopamine (DA) and their principal metabolites, p-hydroxyphenylacetic acid (p-HPAA), m-hydroxyphenylacetic acid (m-HPAA) and homovanillic acid (HVA) were determined in the corpus striatum of Swiss mice at various times after the subcutaneous administration of
P H Yu et al.
Journal of neurochemistry, 45(3), 836-843 (1985-09-01)
An arylamine sulfotransferase (PST-M) from human brain cortex that is involved in the formation of O-sulfate esters of monoamines has been purified 272-fold by ammonium sulfate fractionation, gel filtration, DEAE-cellulose ion-exchange chromatography, chromatofocussing, and hydroxyapatite chromatography. A molecular weight of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service